missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-nitropyridine N-Oxide 98.0+%, TCI America™

Catalog No. p-7119982
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31451G 1 g
B31455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31451G Supplier TCI America Supplier No. B31451G

Chemical Identifiers

CAS 52092-43-0
Molecular Formula C5H3BrN2O3
Molecular Weight (g/mol) 218.99
MDL Number MFCD00160743
InChI Key IRBDHXCXCSFNEQ-UHFFFAOYSA-N
PubChem CID 3904026
IUPAC Name 2-bromo-4-nitropyridin-1-ium-1-olate
SMILES [O-][N+](=O)C1=CC=[N+]([O-])C(Br)=C1

Specifications

Melting Point 146°C
Color Yellow
Boiling Point 430°C
Quantity 1 g
UN Number 2811
Synonym 2-bromo-4-nitropyridine 1-oxide, 2-bromo-4-nitropyridine n-oxide, 2-bromo-4-nitropyridine-n-oxide, 2-bromo-4-nitropyridin-1-ium-1-olate, pyridine, 2-bromo-4-nitro-, 1-oxide, 2-bromo-4-nitropyridine1-oxide, 2-bromo-4-nitropyridine oxide, 2-bromo-4-nitropyridin-1-ol, pubchem5958, acmc-1astu
Formula Weight 218.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-nitropyridine N-Oxide