missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-nitroacetophenone, 95%

Catalog No. AAA1537409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1537409 10 g
AAA1537418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1537409 Supplier Thermo Scientific Chemicals Supplier No. A1537409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 99-81-0 | C8H6BrNO3 | 244.044 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-81-0
Molecular Formula C8H6BrNO3
Molecular Weight (g/mol) 244.044
MDL Number MFCD00007356
InChI Key MBUPVGIGAMCMBT-UHFFFAOYSA-N
Synonym 2-bromo-4'-nitroacetophenone, 2-bromo-1-4-nitrophenyl ethanone, 4-nitrophenacyl bromide, p-nitrophenacyl bromide, 2-bromo-1-4-nitrophenyl ethan-1-one, alpha-bromo-p-nitroacetophenone, ethanone, 2-bromo-1-4-nitrophenyl, 4'-nitrophenacyl bromide, alpha-bromo-4-nitroacetophenone, 2-bromo-4-nitroacetophenone
PubChem CID 66840
IUPAC Name 2-bromo-1-(4-nitrophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)[N+](=O)[O-]

Specifications

Melting Point 95°C to 98°C
Quantity 10 g
UN Number UN1759
Beilstein 393567
Formula Weight 244.05
Percent Purity 95%
Chemical Name or Material 2-Bromo-4'-nitroacetophenone
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 202-789-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only