missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylpyrimidine, 97%

Catalog No. p-7036242
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5450303 1 g
AAH5450306 5 g
AAH5450314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5450303 Supplier Thermo Scientific Chemicals Supplier No. H5450303
Only null left

CAS: 130645-48-6 | C5H5BrN2 | 173.013 g/mol

2-Bromo-4-methylpyrimidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-methylpyrimidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 130645-48-6
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD04037883
InChI Key UZIJEBOLOXOVFY-UHFFFAOYSA-N
PubChem CID 10942928
IUPAC Name 2-bromo-4-methylpyrimidine
SMILES CC1=NC(=NC=C1)Br

Specifications

Melting Point 45°C to 50°C
Quantity 1 g
Synonym 2-bromo-4-methyl-pyrimidine, pyrimidine, 2-bromo-4-methyl, pyrimidine,2-bromo-4-methyl, pubchem7007, acmc-209z1w, 2-bromo-4-methlypyrimidine, 2-bromo-4-methyl pyrimidine, 2-bromo-4-methylpyrimidine
Solubility Information Slightly soluble in water.
Formula Weight 173.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-methylpyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only