missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylpyridine 98.0+%, TCI America™

Catalog No. B21125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21125G 5 g
B211225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21125G Supplier TCI America Supplier No. B21125G
Only null left

Chemical Identifiers

CAS 4926-28-7
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00082590
InChI Key LSZMVESSGLHDJE-UHFFFAOYSA-N
Synonym 2-bromo-4-picoline, 2-bromo-4-methyl-pyridine, 4-methyl-2-bromopyridine, pyridine, 2-bromo-4-methyl, 2 bromo-4-methyl pyridine, 2-bromo-4-methyl pyridine, pubchem1184, acmc-209kfi, 2-bromo4-methylpyridine
PubChem CID 2734087
IUPAC Name 2-bromo-4-methylpyridine
SMILES CC1=CC=NC(Br)=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 172.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-methylpyridine