missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-methylpropiophenone 98.0+%, TCI America™

Catalog No. p-7123745
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47815G 5 g
B478125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47815G Supplier TCI America Supplier No. B47815G
Only null left

Chemical Identifiers

CAS 1451-82-7
Molecular Formula C10H11BrO
Molecular Weight (g/mol) 227.10
MDL Number MFCD11131402
InChI Key OZLUPIIIHOOPNQ-UHFFFAOYNA-N
Synonym 2-bromo-1-p-tolyl propan-1-one, 2-bromo-4'-methylpropiophenone, 2-bromo-1-4-methylphenyl propan-1-one, 2-bromo-4-methylpropiophenone, 2-bromo-1-4-methylphenyl-1-propanone, 1-propanone, 2-bromo-1-4-methylphenyl, 2-bromo-1-p-tolylpropan-1-one, ksc917e0p, 2-bromo-1-p-tolyl-propan-1-one, 4'-methyl-alpha-bromopropiophenone
PubChem CID 2734063
IUPAC Name 2-bromo-1-(4-methylphenyl)propan-1-one
SMILES CC(Br)C(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 80°C
Color White
Boiling Point 162°C
Quantity 5 g
Formula Weight 227.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4′-methylpropiophenone