missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromo-4'-methylacetanilide 98.0+%, TCI America™

Catalog No. B012510G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B012510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B012510G Supplier TCI America Supplier No. B012510G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 614-83-5
Molecular Formula C9H10BrNO
Molecular Weight (g/mol) 228.089
MDL Number MFCD00017781
InChI Key UUDGTWKIIUEVJD-UHFFFAOYSA-N
Synonym n-2-bromo-4-methylphenyl acetamide, 2'-bromo-4'-methylacetanilide, 2-bromo-4-methylacetanilide, acetamide, n-2-bromo-4-methylphenyl, n-acetyl-2-bromo-p-toluidine, acmc-1bdfo, 2'-bromo-p-acetotoluidide, cambridge id 5104354, 4-acetamino-3-bromo-toluene, 2-bromo-4-methyl acetanilide
PubChem CID 69200
IUPAC Name N-(2-bromo-4-methylphenyl)acetamide
SMILES CC1=CC(=C(C=C1)NC(=O)C)Br

Specifications

Melting Point 119°C
Quantity 10 g
Formula Weight 228.09
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Bromo-4′-methylacetanilide