missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-iodopyridine 98.0+%, TCI America™

Catalog No. b51191g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51191G 1 g
B51195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51191G Supplier TCI America Supplier No. B51191G
Only null left

Chemical Identifiers

CAS 100523-96-4
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.894
MDL Number MFCD03085768
InChI Key HPKRNLGLZYOVJS-UHFFFAOYSA-N
PubChem CID 1051520
IUPAC Name 2-bromo-4-iodopyridine
SMILES C1=CN=C(C=C1I)Br

Specifications

Melting Point 64°C
Color White-Yellow
Quantity 1 g
Synonym 2-bromo-4-iodo-pyridine, pyridine, 2-bromo-4-iodo, 4-iodo-2-bromopyridine, pyridine,2-bromo-4-iodo, pubchem5956, acmc-20a0oj, ksc493g9n, abbypharma ap-13-5788
Formula Weight 283.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-iodopyridine