missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-hydroxyacetophenone 98.0+%, TCI America™

Catalog No. B41155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41155G 5 g
B411525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41155G Supplier TCI America Supplier No. B41155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2491-38-5
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00072424
InChI Key LJYOFQHKEWTQRH-UHFFFAOYSA-N
Synonym 4-(Bromoacetyl)phenol, 4-Hydroxyphenacyl Bromide
PubChem CID 4964
IUPAC Name 2-bromo-1-(4-hydroxyphenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)O

Specifications

Melting Point 131°C
Quantity 5 g
UN Number 3261
Formula Weight 215.05
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4′-hydroxyacetophenone
RTECSNumber AM5982000
TSCA No