missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-hexylthiophene 98.0+%, TCI America™

Supplier:  TCI America B39851G

Catalog No. B39851G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Bromo-4-hexylthiophene
Yellow
C10H15BrS
1 g
CCCCCCC1=CSC(=C1)Br
247.194
247.19
Liquid
210705-84-3
144°C
MFCD18413571
IKXKKFNSZQNRFR-UHFFFAOYSA-N
2-bromo-4-hexylthiophene
10916176
≥98.0% (GC)

Chemical Identifiers

210705-84-3
247.194
IKXKKFNSZQNRFR-UHFFFAOYSA-N
2-bromo-4-hexylthiophene
C10H15BrS
MFCD18413571
10916176
CCCCCCC1=CSC(=C1)Br
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions