missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzenesulfonyl chloride, 97%

Catalog No. AAL1971303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1971303 1 g
AAL1971306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1971303 Supplier Thermo Scientific Chemicals Supplier No. L1971303

CAS: 351003-45-7 | C6H3BrClFO2S | 273.5 g/mol

2-Bromo-4-fluorobenzenesulfonyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzenesulfonyl chloride is used as a pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture, bases.

Chemical Identifiers

CAS 351003-45-7
Molecular Formula C6H3BrClFO2S
Molecular Weight (g/mol) 273.5
MDL Number MFCD03094149
InChI Key GJPWPFVLPNTOOL-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorobenzene-1-sulfonyl chloride, 2-bromo-4-fluorobenzenesulphonyl chloride, 2-bromo-4-fluorobenzenesulfonylchloride, 4-fluoro-2-bromobenzenesulfonyl chloride, 2-bromo-4-fluorophenyl chlorosulfone, benzenesulfonyl chloride, 2-bromo-4-fluoro, pubchem8367, acmc-209vnu, 2-bromo-4-fluoro-benzenesulfonyl chloride, ksc221s9l
PubChem CID 2778216
IUPAC Name 2-bromo-4-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1F)Br)S(=O)(=O)Cl

Specifications

Density 1.87
Boiling Point 264°C to 265°C
Refractive Index 1.578
Quantity 1 g
UN Number UN2920
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 273.51
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-fluorobenzenesulfonyl chloride
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only