missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-fluoroacetophenone 97.0+%, TCI America™

Catalog No. p-7119647
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23825G 5 g
B238225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23825G Supplier TCI America Supplier No. B23825G
Only null left

Chemical Identifiers

CAS 403-29-2
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.037
MDL Number MFCD00040830
InChI Key ZJFWCELATJMDNO-UHFFFAOYSA-N
Synonym 2-bromo-4'-fluoroacetophenone, 4-fluorophenacyl bromide, 2-bromo-1-4-fluorophenyl ethanone, 2-bromo-1-4-fluorophenyl ethan-1-one, p-fluorophenacyl bromide, 2'-bromo-4-fluoroacetophenone, ethanone, 2-bromo-1-4-fluorophenyl, bromo-4-fluoroacetophenone, 4-fluorophenacylbromide, 2-bromo-1-4-fluoro-phenyl-ethanone
PubChem CID 96749
IUPAC Name 2-bromo-1-(4-fluorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)F

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 153°C
Quantity 5 g
UN Number 3261
Formula Weight 217.04
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4′-fluoroacetophenone
TSCA No
Recommended Storage Refrigerator