missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-fluoroacetophenone, 98%

Catalog No. AC306290050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC306290050 5 g Glass bottle
AC306290250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC306290050 Supplier Thermo Scientific Chemicals Supplier No. 306290050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 403-29-2 | C8H6BrFO | 217.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 403-29-2
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.04
MDL Number MFCD00040830
InChI Key ZJFWCELATJMDNO-UHFFFAOYSA-N
Synonym 2-bromo-4'-fluoroacetophenone, 4-fluorophenacyl bromide, 2-bromo-1-4-fluorophenyl ethanone, 2-bromo-1-4-fluorophenyl ethan-1-one, p-fluorophenacyl bromide, 2'-bromo-4-fluoroacetophenone, ethanone, 2-bromo-1-4-fluorophenyl, bromo-4-fluoroacetophenone, 4-fluorophenacylbromide, 2-bromo-1-4-fluoro-phenyl-ethanone
PubChem CID 96749
IUPAC Name 2-bromo-1-(4-fluorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 46.0°C to 49.0°C
Color Gray to White
Boiling Point 150.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4COCH2Br
Quantity 5 g
Formula Weight 217.04
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Bromo-4'-fluoroacetophenone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-955-1

RUO – Research Use Only