Learn More
2-Bromo-4'-fluoroacetophenone, 97%
CAS: 403-29-2 | C8H6BrFO | 217.037 g/mol
Supplier: Thermo Scientific Chemicals A1531906
Description
2-Bromo-4'-fluoroacetophenone is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-4′-fluoroacetophenone is used as an organic chemical synthesis intermediate.
Solubility
Soluble in methanol.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications
| 2-Bromo-4'-fluoroacetophenone | |
| 45°C to 49°C | |
| >110°C (230°F) | |
| MFCD00040830 | |
| 637863 | |
| Soluble in methanol. | |
| C1=CC(=CC=C1C(=O)CBr)F | |
| 217.037 | |
| 217.04 |
| 403-29-2 | |
| 150°C (12 mmHg) | |
| C8H6BrFO | |
| 5 g | |
| 2-bromo-4'-fluoroacetophenone, 4-fluorophenacyl bromide, 2-bromo-1-4-fluorophenyl ethanone, 2-bromo-1-4-fluorophenyl ethan-1-one, p-fluorophenacyl bromide, 2'-bromo-4-fluoroacetophenone, ethanone, 2-bromo-1-4-fluorophenyl, bromo-4-fluoroacetophenone, 4-fluorophenacylbromide, 2-bromo-1-4-fluoro-phenyl-ethanone | |
| ZJFWCELATJMDNO-UHFFFAOYSA-N | |
| 2-bromo-1-(4-fluorophenyl)ethanone | |
| 96749 | |
| 97% |
Chemical Identifiers
| 403-29-2 | |
| 217.037 | |
| ZJFWCELATJMDNO-UHFFFAOYSA-N | |
| 96749 | |
| C1=CC(=CC=C1C(=O)CBr)F |
| C8H6BrFO | |
| MFCD00040830 | |
| 2-bromo-4'-fluoroacetophenone, 4-fluorophenacyl bromide, 2-bromo-1-4-fluorophenyl ethanone, 2-bromo-1-4-fluorophenyl ethan-1-one, p-fluorophenacyl bromide, 2'-bromo-4-fluoroacetophenone, ethanone, 2-bromo-1-4-fluorophenyl, bromo-4-fluoroacetophenone, 4-fluorophenacylbromide, 2-bromo-1-4-fluoro-phenyl-ethanone | |
| 2-bromo-1-(4-fluorophenyl)ethanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 206-955-1
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only