missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-fluoroacetophenone, 97%

Catalog No. AAA1531903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1531903 1 g
AAA1531906 5 g
AAA1531914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1531903 Supplier Thermo Scientific Chemicals Supplier No. A1531903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 403-29-2 | C8H6BrFO | 217.037 g/mol

2-Bromo-4'-fluoroacetophenone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-fluoroacetophenone is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Moisture sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 403-29-2
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.037
MDL Number MFCD00040830
InChI Key ZJFWCELATJMDNO-UHFFFAOYSA-N
Synonym 2-bromo-4'-fluoroacetophenone, 4-fluorophenacyl bromide, 2-bromo-1-4-fluorophenyl ethanone, 2-bromo-1-4-fluorophenyl ethan-1-one, p-fluorophenacyl bromide, 2'-bromo-4-fluoroacetophenone, ethanone, 2-bromo-1-4-fluorophenyl, bromo-4-fluoroacetophenone, 4-fluorophenacylbromide, 2-bromo-1-4-fluoro-phenyl-ethanone
PubChem CID 96749
IUPAC Name 2-bromo-1-(4-fluorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)F

Specifications

Melting Point 45°C to 49°C
Boiling Point 150°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 637863
Solubility Information Soluble in methanol.
Formula Weight 217.04
Percent Purity 97%
Chemical Name or Material 2-Bromo-4'-fluoroacetophenone
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 206-955-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only