missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-(diethylamino)acetophenone, 98%

Catalog No. AAL1333703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1333703 1 g
AAL1333706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1333703 Supplier Thermo Scientific Chemicals Supplier No. L1333703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 207986-25-2 | C12H16BrNO | 270.17 g/mol

2-Bromo-4'-(diethylamino)acetophenone, is used as a pharmaceutical intermediate. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-(diethylamino)acetophenone, is used as a pharmaceutical intermediate. It is also used as a primary and secondary intermediate.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 207986-25-2
Molecular Formula C12H16BrNO
Molecular Weight (g/mol) 270.17
MDL Number MFCD00151798
InChI Key ALTMCDJIFXTEPV-UHFFFAOYSA-N
Synonym 2-bromo-1-4-diethylamino phenyl ethanone, alpha-bromo-4-diethylamino acetophenone, 2-bromo-4'-diethylamino acetophenone, 2-bromo-1-4-diethylamino phenyl ethan-1-one, 4-diethylaminophenacyl bromide, n,n-diethyl-4-bromoacetyl aniline, 4-n,n-diethylamino phenacyl bromide, .alpha.-bromo-4-diethylaminoacetophenone, 2-bromo-1-4-diethylaminophenyl ethanone
PubChem CID 2795030
IUPAC Name 2-bromo-1-[4-(diethylamino)phenyl]ethanone
SMILES CCN(CC)C1=CC=C(C=C1)C(=O)CBr

Specifications

Melting Point 50°C to 52°C
Quantity 1 g
Beilstein 7921481
Solubility Information It is insoluble in water.
Formula Weight 270.18
Percent Purity 98%
Chemical Name or Material 2-Bromo-4'-(diethylamino)acetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only