missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chlorotoluene, 98%

Catalog No. p-7028171
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2532914 25 g
AAB2532922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2532914 Supplier Thermo Scientific Chemicals Supplier No. B2532914

CAS: 27139-97-5 | C7H6BrCl | 205.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27139-97-5
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00060650
InChI Key CSUUXPHPCXHYGY-UHFFFAOYSA-N
Synonym 2-bromo-4-chlorotoluene, benzene, 2-bromo-4-chloro-1-methyl, 4-chloro-2-bromotoluene, toluene, 2-bromo-4-chloro, p-chlor brom toluol, pubchem3599, acmc-209gvg, 2-bromo-4-chloro toluene, 2-bromo-4-chloro-toluene, ksc493k9b
PubChem CID 609898
IUPAC Name 2-bromo-4-chloro-1-methylbenzene
SMILES CC1=C(Br)C=C(Cl)C=C1

Specifications

Quantity 25 g
Formula Weight 205.48
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-chlorotoluene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only