missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-chloropyridine 98.0+%, TCI America™
$55.84 - $134.75
Chemical Identifiers
| CAS | 22918-01-0 |
|---|---|
| Molecular Formula | C5H3BrClN |
| Molecular Weight (g/mol) | 192.44 |
| MDL Number | MFCD03840757 |
| InChI Key | SURKZMFXICWLHU-UHFFFAOYSA-N |
| Synonym | 2-bromo-4-chloro-pyridine, pyridine, 2-bromo-4-chloro, pubchem13533, acmc-1cled, 2-brom-4-chlorpyridin;, 2-bromo-4 chloropyridine, 2-bromo-4-chloro pyridine, 2-bromo-4-chloropyridine;, 2-bromo-4-pyridyl chloride, 2-bromo-4-pyridyl chloride; |
| PubChem CID | 2762836 |
| IUPAC Name | 2-bromo-4-chloropyridine |
| SMILES | ClC1=CC=NC(Br)=C1 |
Chemical Identifiers
| 22918-01-0 | |
| 192.44 | |
| SURKZMFXICWLHU-UHFFFAOYSA-N | |
| 2762836 | |
| ClC1=CC=NC(Br)=C1 |
| C5H3BrClN | |
| MFCD03840757 | |
| 2-bromo-4-chloro-pyridine, pyridine, 2-bromo-4-chloro, pubchem13533, acmc-1cled, 2-brom-4-chlorpyridin;, 2-bromo-4 chloropyridine, 2-bromo-4-chloro pyridine, 2-bromo-4-chloropyridine;, 2-bromo-4-pyridyl chloride, 2-bromo-4-pyridyl chloride; | |
| 2-bromo-4-chloropyridine |
Specifications
| 22918-01-0 | |
| White | |
| C5H3BrClN | |
| 1 g | |
| SURKZMFXICWLHU-UHFFFAOYSA-N | |
| 2-bromo-4-chloropyridine | |
| 2762836 | |
| ≥98.0% (GC) | |
| 2-Bromo-4-chloropyridine |
| 27°C | |
| 85°C | |
| MFCD03840757 | |
| 2-bromo-4-chloro-pyridine, pyridine, 2-bromo-4-chloro, pubchem13533, acmc-1cled, 2-brom-4-chlorpyridin;, 2-bromo-4 chloropyridine, 2-bromo-4-chloro pyridine, 2-bromo-4-chloropyridine;, 2-bromo-4-pyridyl chloride, 2-bromo-4-pyridyl chloride; | |
| ClC1=CC=NC(Br)=C1 | |
| 192.44 | |
| 192.44 | |
| Crystalline Powder |
Safety and Handling
TSCA : No