missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloropyridine, 97%

Catalog No. AAH3369803
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH3369803 1 g
AAH3369806 5 g
2 options

Catalog No. AAH3369803

Supplier: Thermo Scientific Chemicals H3369803

Only null left

CAS: 22918-01-0 | C5H3BrClN | 192.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22918-01-0
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD03840757
InChI Key SURKZMFXICWLHU-UHFFFAOYSA-N
Synonym 2-bromo-4-chloro-pyridine, pyridine, 2-bromo-4-chloro, pubchem13533, acmc-1cled, 2-brom-4-chlorpyridin;, 2-bromo-4 chloropyridine, 2-bromo-4-chloro pyridine, 2-bromo-4-chloropyridine;, 2-bromo-4-pyridyl chloride, 2-bromo-4-pyridyl chloride;
PubChem CID 2762836
IUPAC Name 2-bromo-4-chloropyridine
SMILES ClC1=CC=NC(Br)=C1

Specifications

Melting Point <30°C
Quantity 1 g
Formula Weight 192.44
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-chloropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only