Learn More
Chemical Identifiers
| CAS | 873-38-1 |
|---|---|
| Molecular Formula | C6H5BrClN |
| Molecular Weight (g/mol) | 206.47 |
| MDL Number | MFCD00041313 |
| InChI Key | SYTBIFURTZACKR-UHFFFAOYSA-N |
| Synonym | benzenamine, 2-bromo-4-chloro, 1-amino-2-bromo-4-chlorobenzene, 2-bromo-4-chloro-phenylamine, 2-bromo-4-chloro aniline, 2-bromo-4-chlorophenylamine, pubchem3576, acmc-1bkdm, 2-bromo-4-chloro-aniline, 4-chloro-2-bromoaniline, 2-bromo-4-chloroaniline |
| PubChem CID | 70110 |
| IUPAC Name | 2-bromo-4-chloroaniline |
| SMILES | C1=CC(=C(C=C1Cl)Br)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC393470010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C6H5BrClN | |
| MFCD00041313 | |
| benzenamine, 2-bromo-4-chloro, 1-amino-2-bromo-4-chlorobenzene, 2-bromo-4-chloro-phenylamine, 2-bromo-4-chloro aniline, 2-bromo-4-chlorophenylamine, pubchem3576, acmc-1bkdm, 2-bromo-4-chloro-aniline, 4-chloro-2-bromoaniline, 2-bromo-4-chloroaniline | |
| 2-bromo-4-chloroaniline |
Specifications
| 873-38-1 | |
| 100.0 | |
| Authentic | |
| C6H5BrClN | |
| MFCD00041313 | |
| SYTBIFURTZACKR-UHFFFAOYSA-N | |
| 2-bromo-4-chloroaniline | |
| 70110 | |
| 98% |
| 97.5 | |
| 127°C (20.0mmHg) | |
| 97.5% min. (GC) | |
| ClC6H3(Br)NH2 | |
| benzenamine, 2-bromo-4-chloro, 1-amino-2-bromo-4-chlorobenzene, 2-bromo-4-chloro-phenylamine, 2-bromo-4-chloro aniline, 2-bromo-4-chlorophenylamine, pubchem3576, acmc-1bkdm, 2-bromo-4-chloro-aniline, 4-chloro-2-bromoaniline, 2-bromo-4-chloroaniline | |
| C1=CC(=C(C=C1Cl)Br)N | |
| 206.47 | |
| 206.47 | |
| 2-Bromo-4-chloroaniline |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 212-837-
RUO – Research Use Only