missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-diphenylpyrimidine 97.0+%, TCI America™

Catalog No. p-7123676
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4662200MG 200 mg
B46621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4662200MG Supplier TCI America Supplier No. B4662200MG
Only null left

Chemical Identifiers

CAS 56181-49-8
Molecular Formula C16H11BrN2
Molecular Weight (g/mol) 311.182
MDL Number MFCD00234884
InChI Key GPGIIKKUKINTGW-UHFFFAOYSA-N
PubChem CID 1712519
IUPAC Name 2-bromo-4,6-diphenylpyrimidine
SMILES C1=CC=C(C=C1)C2=CC(=NC(=N2)Br)C3=CC=CC=C3

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 200 mg
Formula Weight 311.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4,6-diphenylpyrimidine