missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™
$231.14 - $697.16
Chemical Identifiers
| CAS | 80984-79-8 |
|---|---|
| Molecular Formula | C15H10BrN3 |
| Molecular Weight (g/mol) | 312.17 |
| MDL Number | MFCD10687147 |
| InChI Key | PTPGZCQGDXUUAH-UHFFFAOYSA-N |
| PubChem CID | 27281662 |
| IUPAC Name | 2-bromo-4,6-diphenyl-1,3,5-triazine |
| SMILES | C1=CC=C(C=C1)C2=NC(=NC(=N2)Br)C3=CC=CC=C3 |
Chemical Identifiers
| 80984-79-8 | |
| 312.17 | |
| PTPGZCQGDXUUAH-UHFFFAOYSA-N | |
| 2-bromo-4,6-diphenyl-1,3,5-triazine |
| C15H10BrN3 | |
| MFCD10687147 | |
| 27281662 | |
| C1=CC=C(C=C1)C2=NC(=NC(=N2)Br)C3=CC=CC=C3 |
Specifications
| 80984-79-8 | |
| C15H10BrN3 | |
| 1 g | |
| C1=CC=C(C=C1)C2=NC(=NC(=N2)Br)C3=CC=CC=C3 | |
| 312.17 | |
| 312.17 | |
| Crystalline Powder |
| 156°C | |
| MFCD10687147 | |
| PTPGZCQGDXUUAH-UHFFFAOYSA-N | |
| 2-bromo-4,6-diphenyl-1,3,5-triazine | |
| 27281662 | |
| ≥98.0% (GC) | |
| 2-Bromo-4,6-diphenyl-1,3,5-triazine |
Safety and Handling
TSCA : No