missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™

Catalog No. B49391G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49391G 1 g
B49395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49391G Supplier TCI America Supplier No. B49391G
Only null left

Chemical Identifiers

CAS 80984-79-8
Molecular Formula C15H10BrN3
Molecular Weight (g/mol) 312.17
MDL Number MFCD10687147
InChI Key PTPGZCQGDXUUAH-UHFFFAOYSA-N
PubChem CID 27281662
IUPAC Name 2-bromo-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=NC(=NC(=N2)Br)C3=CC=CC=C3

Specifications

Melting Point 156°C
Quantity 1 g
Formula Weight 312.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4,6-diphenyl-1,3,5-triazine