missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. B31791G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31791G 1 g
B31795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31791G Supplier TCI America Supplier No. B31791G

Chemical Identifiers

CAS 175205-82-0
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 225.996
MDL Number MFCD00153084
InChI Key OFGSIPQYQUVVPL-UHFFFAOYSA-N
Synonym 2-bromo-3-trifluoromethyl pyridine, 2-bromo-3-trifluorormethyl pyridine, pyridine, 2-bromo-3-trifluoromethyl, 2-bromo-3-trifluormethyl-pyridine, pubchem2999, acmc-1bx8n, ksc174i2d, tpc-py049, buttpark 27\06-85
PubChem CID 5193820
IUPAC Name 2-bromo-3-(trifluoromethyl)pyridine
SMILES C1=CC(=C(N=C1)Br)C(F)(F)F

Specifications

Melting Point 40°C
Boiling Point 69°C
Quantity 1 g
UN Number 2811
Formula Weight 226.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-(trifluoromethyl)pyridine