missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-tetradecylthiophene 95.0+%, TCI America™

Catalog No. B54081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54081G 1 g
B54085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54081G Supplier TCI America Supplier No. B54081G
Only null left

Chemical Identifiers

CAS 500199-09-7
Molecular Formula C18H31BrS
Molecular Weight (g/mol) 359.41
InChI Key GOUPSYVDGUEWMF-UHFFFAOYSA-N
PubChem CID 67293590
IUPAC Name 2-bromo-3-tetradecylthiophene
SMILES CCCCCCCCCCCCCCC1=C(SC=C1)Br

Specifications

Quantity 1 g
Formula Weight 359.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-tetradecylthiophene