missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-pyridinecarboxaldehyde, 96%

Catalog No. AC399670010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399670010 1 g Glass bottle
AC399670050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399670010 Supplier Thermo Scientific Chemicals Supplier No. 399670010

CAS: 128071-75-0 | C6H4BrNO | 186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 128071-75-0
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186
MDL Number MFCD04966945
InChI Key GNFWMEFWZWXLIN-UHFFFAOYSA-N
Synonym 2-bromonicotinaldehyde, 2-bromo-3-formylpyridine, 2-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-bromo, pubchem5057, acmc-1bypv, 2-bromo-3-formyl pyridine, bromo-3-pyridine carbaldehyde, bromo-3-pyridinecarboxaldehyde
PubChem CID 4714951
IUPAC Name 2-bromopyridine-3-carbaldehyde
SMILES C1=CC(=C(N=C1)Br)C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 70.0°C to 74.0°C
Boiling Point 100.0°C (3.0 mmHg)
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 186
Percent Purity 96%
Chemical Name or Material 2-Bromo-3-pyridinecarboxaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only