missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-pentanone 95.0+%, TCI America™

Catalog No. b464825g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B464825G 25 g
B46485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B464825G Supplier TCI America Supplier No. B464825G
Only null left

Chemical Identifiers

CAS 815-52-1
Molecular Formula C5H9BrO
Molecular Weight (g/mol) 165.03
MDL Number MFCD11036312
InChI Key VUDTYIUNUSPULX-UHFFFAOYSA-N
PubChem CID 545092
IUPAC Name 2-bromopentan-3-one
SMILES CCC(=O)C(C)Br

Specifications

Color Yellow
Boiling Point 55°C
Quantity 25 g
UN Number 1224
Formula Weight 165.03
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-pentanone
TSCA No
Recommended Storage Refrigerator