missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-nitropyridine 98.0+%, TCI America™

Catalog No. B41935G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41935G 5 g
B41931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41935G Supplier TCI America Supplier No. B41935G

Chemical Identifiers

CAS 19755-53-4
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 203.00
MDL Number MFCD00955613
InChI Key ZCCUFLITCDHRMG-UHFFFAOYSA-N
Synonym 2-bromo-3-nitro-pyridine, pyridine, 2-bromo-3-nitro, pyridine, 2-bromonitro, pubchem5955, acmc-1bt4y, 2-bromo-3-nitro pyridine, #, ksc490s2t, 2-bromo-3-nitropyridine
PubChem CID 555054
IUPAC Name 2-bromo-3-nitropyridine
SMILES [O-][N+](=O)C1=CC=CN=C1Br

Specifications

Melting Point 124°C
Color Yellow
Quantity 5 g
Formula Weight 203.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-nitropyridine