missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylpyridine 97.0+%, TCI America™

Catalog No. p-7119335
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18945G 5 g
B189425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18945G Supplier TCI America Supplier No. B18945G

Chemical Identifiers

CAS 3430-17-9
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00239380
InChI Key PZSISEFPCYMBDL-UHFFFAOYSA-N
Synonym 2-bromo-3-picoline, 2-bromo-3-methylpyridin, 2-bromo-3-methyl pyridine, 2-bromo-3-methyl-pyridine, 3-methyl-2-bromopyridine, pyridine, 2-bromo-3-methyl, 2-brom-3-methylpyridin, pubchem9208, 2-bromo-3-mehtylpyridine
PubChem CID 220832
IUPAC Name 2-bromo-3-methylpyridine
SMILES CC1=CC=CN=C1Br

Specifications

Color Yellow
Boiling Point 219°C
Quantity 5 g
Formula Weight 172.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-methylpyridine