missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylpyridine, 98+%

Catalog No. AAL1952206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1952206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1952206 Supplier Thermo Scientific Chemicals Supplier No. L1952206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3430-17-9 | C6H6BrN | 172.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3430-17-9
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00239380
InChI Key PZSISEFPCYMBDL-UHFFFAOYSA-N
Synonym 2-bromo-3-picoline, 2-bromo-3-methylpyridin, 2-bromo-3-methyl pyridine, 2-bromo-3-methyl-pyridine, 3-methyl-2-bromopyridine, pyridine, 2-bromo-3-methyl, 2-brom-3-methylpyridin, pubchem9208, 2-bromo-3-mehtylpyridine
PubChem CID 220832
IUPAC Name 2-bromo-3-methylpyridine
SMILES CC1=CC=CN=C1Br

Specifications

Density 1.544
Boiling Point 218°C to 219°C
Flash Point >110°C (230°F)
Refractive Index 1.5695
Quantity 5 g
Beilstein 107896
Formula Weight 172.03
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-3-methylpyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only