missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylpyridine, 95%, Thermo Scientific™

Chemical Identifiers

CAS 3430-17-9
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00239380
InChI Key PZSISEFPCYMBDL-UHFFFAOYSA-N
Synonym 2-bromo-3-picoline, 2-bromo-3-methylpyridin, 2-bromo-3-methyl pyridine, 2-bromo-3-methyl-pyridine, 3-methyl-2-bromopyridine, pyridine, 2-bromo-3-methyl, 2-brom-3-methylpyridin, pubchem9208, 2-bromo-3-mehtylpyridine
PubChem CID 220832
IUPAC Name 2-bromo-3-methylpyridine
SMILES CC1=CC=CN=C1Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC357660050
SDS View Documents
Thermo Scientific Chemicals
357660050
5mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3430-17-9
172.03
PZSISEFPCYMBDL-UHFFFAOYSA-N
220832
CC1=CC=CN=C1Br
C6H6BrN
MFCD00239380
2-bromo-3-picoline, 2-bromo-3-methylpyridin, 2-bromo-3-methyl pyridine, 2-bromo-3-methyl-pyridine, 3-methyl-2-bromopyridine, pyridine, 2-bromo-3-methyl, 2-brom-3-methylpyridin, pubchem9208, 2-bromo-3-mehtylpyridine
2-bromo-3-methylpyridine

Specifications

3430-17-9
100.0
>110°C
94% min. (GC)
C6H6BrN
MFCD00239380
1.544
PZSISEFPCYMBDL-UHFFFAOYSA-N
2-bromo-3-methylpyridine
220832
95%
94.0
1.5440g/mL
Authentic
Glass bottle
1.5650 to 1.57
5mL
2-bromo-3-picoline, 2-bromo-3-methylpyridin, 2-bromo-3-methyl pyridine, 2-bromo-3-methyl-pyridine, 3-methyl-2-bromopyridine, pyridine, 2-bromo-3-methyl, 2-brom-3-methylpyridin, pubchem9208, 2-bromo-3-mehtylpyridine
CC1=CC=CN=C1Br
172.03
172.02
2-Bromo-3-methylpyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only