missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methyl-5-nitropyridine 98.0+%, TCI America™

Catalog No. p-7119764
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B27985G Supplier TCI America Supplier No. B27985G
Only null left

Chemical Identifiers

CAS 23132-21-0
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
InChI Key FZIQHPKXSLHGBZ-UHFFFAOYSA-N
Synonym 2-bromo-5-nitro-3-picoline, 2-bromo-3-methyl-5-nitro-pyridine, pyridine, 2-bromo-3-methyl-5-nitro, 2-hydroxy-5-nitro-3-picoline 2-hydroxy-3-methyl-5-nitropyridine, pubchem10608, acmc-209g1c, ksc496c3r, fziqhpkxslhgbz-uhfffaoysa, 2-bromo-3-methyl-5-nitro pyridine, 2-bromanyl-3-methyl-5-nitro-pyridine
PubChem CID 5200466
IUPAC Name 2-bromo-3-methyl-5-nitropyridine
SMILES CC1=CC(=CN=C1Br)[N+](=O)[O-]

Specifications

Melting Point 58°C
Quantity 5 g
Formula Weight 217.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-methyl-5-nitropyridine