missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-methoxypyridine 98.0+%, TCI America™
Supplier: TCI America B386125G
Specifications
| 2-Bromo-3-methoxypyridine | |
| 47°C | |
| C6H6BrNO | |
| 25 g | |
| PDOWLYNSFYZIQX-UHFFFAOYSA-N | |
| 2-bromo-3-methoxypyridine | |
| 90364 | |
| ≥98.0% (GC) |
| 24100-18-3 | |
| White-Yellow | |
| MFCD01570896 | |
| 2-bromo-3-methoxy-pyridine, pyridine, 2-bromo-3-methoxy, 2-bromo-3-methoxy pyridine, pubchem5916, acmc-1cefu, ksc552s2b, #, pyridine,2-bromo-3-methoxy, 2-bromo-3-methoxypyridine, abbypharma ap-17-5948 | |
| COC1=C(N=CC=C1)Br | |
| 188.024 | |
| 188.02 | |
| Crystalline Powder |
Chemical Identifiers
| 24100-18-3 | |
| 188.024 | |
| PDOWLYNSFYZIQX-UHFFFAOYSA-N | |
| 2-bromo-3-methoxypyridine |
| C6H6BrNO | |
| MFCD01570896 | |
| 90364 | |
| COC1=C(N=CC=C1)Br |
Safety and Handling
TSCA : No