missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methoxypyridine 98.0+%, TCI America™

Catalog No. B38615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38615G 5 g
B386125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38615G Supplier TCI America Supplier No. B38615G
Only null left

Chemical Identifiers

CAS 24100-18-3
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD01570896
InChI Key PDOWLYNSFYZIQX-UHFFFAOYSA-N
PubChem CID 90364
IUPAC Name 2-bromo-3-methoxypyridine
SMILES COC1=C(N=CC=C1)Br

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 5 g
Synonym 2-bromo-3-methoxy-pyridine, pyridine, 2-bromo-3-methoxy, 2-bromo-3-methoxy pyridine, pubchem5916, acmc-1cefu, ksc552s2b, #, pyridine,2-bromo-3-methoxy, 2-bromo-3-methoxypyridine, abbypharma ap-17-5948
Formula Weight 188.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-methoxypyridine