missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-hexylthiophene 98.0+%, TCI America™
$218.10 - $671.06
Chemical Identifiers
| CAS | 69249-61-2 |
|---|---|
| Molecular Formula | C10H15BrS |
| Molecular Weight (g/mol) | 247.194 |
| MDL Number | MFCD09907959 |
| InChI Key | XQJNXCHDODCAJF-UHFFFAOYSA-N |
| Synonym | 2-bromo-3-hexyl-thiophene, thiophene, 2-bromo-3-hexyl, acmc-209o6r, 2-bromo-3-hexyl thiophene, 2-bromo-3-n-hexylthiophene, amth047, 2-bromanyl-3-hexyl-thiophene, ksc491i5d, 2-bromo-3-hexylthiophene |
| PubChem CID | 10960141 |
| IUPAC Name | 2-bromo-3-hexylthiophene |
| SMILES | CCCCCCC1=C(SC=C1)Br |
Chemical Identifiers
| 69249-61-2 | |
| 247.194 | |
| XQJNXCHDODCAJF-UHFFFAOYSA-N | |
| 10960141 | |
| CCCCCCC1=C(SC=C1)Br |
| C10H15BrS | |
| MFCD09907959 | |
| 2-bromo-3-hexyl-thiophene, thiophene, 2-bromo-3-hexyl, acmc-209o6r, 2-bromo-3-hexyl thiophene, 2-bromo-3-n-hexylthiophene, amth047, 2-bromanyl-3-hexyl-thiophene, ksc491i5d, 2-bromo-3-hexylthiophene | |
| 2-bromo-3-hexylthiophene |
Specifications
| 69249-61-2 | |
| 75°C | |
| MFCD09907959 | |
| 2-bromo-3-hexyl-thiophene, thiophene, 2-bromo-3-hexyl, acmc-209o6r, 2-bromo-3-hexyl thiophene, 2-bromo-3-n-hexylthiophene, amth047, 2-bromanyl-3-hexyl-thiophene, ksc491i5d, 2-bromo-3-hexylthiophene | |
| CCCCCCC1=C(SC=C1)Br | |
| 247.194 | |
| 247.19 | |
| Liquid |
| Yellow | |
| C10H15BrS | |
| 5 g | |
| XQJNXCHDODCAJF-UHFFFAOYSA-N | |
| 2-bromo-3-hexylthiophene | |
| 10960141 | |
| ≥98.0% (GC) | |
| 2-Bromo-3-hexylthiophene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator