missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluoropyridine 98.0+%, TCI America™

Catalog No. p-7123087
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36661G 1 g
B36665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36661G Supplier TCI America Supplier No. B36661G
Only null left

Chemical Identifiers

CAS 40273-45-8
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
MDL Number MFCD04114128
InChI Key IFGLECYAEGYLSJ-UHFFFAOYSA-N
Synonym 2-bromo-3-fluoro-pyridine, pyridine, 2-bromo-3-fluoro, abbypharma ap-17-5102, pubchem3002, 2-bromo-fluoropyridine, bromo-3-fluoro pyrdine, 2-bromo-3-fluoropyrdine, acmc-1am6d, ksc235k8b
PubChem CID 2762803
IUPAC Name 2-bromo-3-fluoropyridine
SMILES C1=CC(=C(N=C1)Br)F

Specifications

Color Yellow
Boiling Point 62°C
Quantity 1 g
UN Number 1993
Formula Weight 175.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-fluoropyridine