missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluoropyridine, 97%

Catalog No. p-7031047
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3198103 1 g
AAH3198106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3198103 Supplier Thermo Scientific Chemicals Supplier No. H3198103

CAS: 40273-45-8 | C5H3BrFN | 175.988 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40273-45-8
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
MDL Number MFCD04114128
InChI Key IFGLECYAEGYLSJ-UHFFFAOYSA-N
Synonym 2-bromo-3-fluoro-pyridine, pyridine, 2-bromo-3-fluoro, abbypharma ap-17-5102, pubchem3002, 2-bromo-fluoropyridine, bromo-3-fluoro pyrdine, 2-bromo-3-fluoropyrdine, acmc-1am6d, ksc235k8b
PubChem CID 2762803
IUPAC Name 2-bromo-3-fluoropyridine
SMILES C1=CC(=C(N=C1)Br)F

Specifications

Refractive Index 1.545
Quantity 1 g
UN Number UN2810
Formula Weight 175.99
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-fluoropyridine
Hazard Category H227-H301+H311-H315-H319-H332-H335
Hazard Statement GHS H Statement
H301-H311-H227-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only