missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluoropyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH6429006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6429006 5 g
AAH6429003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6429006 Supplier Thermo Scientific Chemicals Supplier No. H6429006

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1211530-89-0
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 220.00
MDL Number MFCD13185796
InChI Key DHFIVEUMGCJILQ-UHFFFAOYSA-N
Synonym 2-bromo-3-fluoroisonicotinic acid, 2-bromo-3-fluoroisonicotinicacid, 2-bromo-3-fluoro-4-picolic acid, 2-bromo-3-fluoro-4-pyridinecarboxylic acid, 2-bromo-3-fluoropyridin-4-carboxylicacid, 2-bromo-3-fluoropyridin-4-carboxylic acid
PubChem CID 70680697
IUPAC Name 2-bromo-3-fluoropyridine-4-carboxylic acid
SMILES OC(=O)C1=C(F)C(Br)=NC=C1

Specifications

Quantity 5 g
Formula Weight 220
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-3-fluoropyridine-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only