missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluoropropionic acid, 96%, Thermo Scientific Chemicals

Catalog No. p-7030844
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3171803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3171803 Supplier Thermo Scientific Chemicals Supplier No. H3171803

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16652-36-1
Molecular Formula C3H4BrFO2
Molecular Weight (g/mol) 170.97
MDL Number MFCD09800640
InChI Key FBPUCVAAEPPEMG-UHFFFAOYNA-N
Synonym 2-bromo-3-fluoropropionic acid, acmc-20ao2z, 3-fluoro-2-bromo-propionic acid
PubChem CID 316511
IUPAC Name 2-bromo-3-fluoropropanoic acid
SMILES OC(=O)C(Br)CF

Specifications

Boiling Point 62°C to 63°C (0.2 mmHg)
Refractive Index 1.4655
Quantity 1 g
UN Number UN3265
Formula Weight 170.97
Percent Purity 96%
Chemical Name or Material 2-Bromo-3-fluoropropionic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only