missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-dodecylthiophene 97.0+%, TCI America™

Catalog No. p-7123107
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B36911G 1 g
B36915G 5 g
2 options

Catalog No. B36911G

Supplier: TCI America B36911G

Only null left

Chemical Identifiers

CAS 139100-06-4
Molecular Formula C16H27BrS
Molecular Weight (g/mol) 331.36
MDL Number MFCD10000891
InChI Key IMILVTHOOISGRW-UHFFFAOYSA-N
Synonym 2-Bromo-3-laurylthiophene
PubChem CID 10426816
IUPAC Name 2-bromo-3-dodecylthiophene
SMILES CCCCCCCCCCCCC1=C(Br)SC=C1

Specifications

Boiling Point 165°C
Quantity 1 g
Formula Weight 331.36
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-dodecylthiophene