missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-decylthiophene 97.0+%, TCI America™

Catalog No. B44555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44555G 5 g
B445525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44555G Supplier TCI America Supplier No. B44555G
Only null left

Chemical Identifiers

CAS 144012-09-9
Molecular Formula C14H23BrS
Molecular Weight (g/mol) 303.30
MDL Number MFCD16619251
InChI Key YCWPDFSNGOICAB-UHFFFAOYSA-N
PubChem CID 46911875
IUPAC Name 2-bromo-3-decylthiophene
SMILES CCCCCCCCCCC1=C(Br)SC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 303.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-decylthiophene