missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-cyclohexylthiophene 95.0+%, TCI America™

Catalog No. B54205G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54205G 5 g
B54201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54205G Supplier TCI America Supplier No. B54205G
Only null left

Chemical Identifiers

CAS 241477-71-4
Molecular Formula C10H13BrS

Specifications

Color Yellow
Quantity 5 g
Formula Weight 245.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-cyclohexylthiophene
TSCA No
Recommended Storage Refrigerator