missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-chloropyridine 98.0+%, TCI America™
$40.43 - $157.87
Chemical Identifiers
| CAS | 96424-68-9 |
|---|---|
| Molecular Formula | C5H3BrClN |
| Molecular Weight (g/mol) | 192.44 |
| MDL Number | MFCD04039344 |
| InChI Key | GOHBBINNYAWQGO-UHFFFAOYSA-N |
| Synonym | pyridine, 2-bromo-3-chloro, 3-chloro-2-bromopyridine, 2-bromo-3-chloro-pyridine, chloro bromo pyridine, pubchem5776, 2-bromo-3-pyridyl chloride, ksc486m1b, 2-bromo-3-chloropyridine, abbypharma ap-18-5389 |
| PubChem CID | 7016326 |
| IUPAC Name | 2-bromo-3-chloropyridine |
| SMILES | C1=CC(=C(N=C1)Br)Cl |
Chemical Identifiers
| 96424-68-9 | |
| 192.44 | |
| GOHBBINNYAWQGO-UHFFFAOYSA-N | |
| 7016326 | |
| C1=CC(=C(N=C1)Br)Cl |
| C5H3BrClN | |
| MFCD04039344 | |
| pyridine, 2-bromo-3-chloro, 3-chloro-2-bromopyridine, 2-bromo-3-chloro-pyridine, chloro bromo pyridine, pubchem5776, 2-bromo-3-pyridyl chloride, ksc486m1b, 2-bromo-3-chloropyridine, abbypharma ap-18-5389 | |
| 2-bromo-3-chloropyridine |
Specifications
| 96424-68-9 | |
| White-Yellow | |
| MFCD04039344 | |
| pyridine, 2-bromo-3-chloro, 3-chloro-2-bromopyridine, 2-bromo-3-chloro-pyridine, chloro bromo pyridine, pubchem5776, 2-bromo-3-pyridyl chloride, ksc486m1b, 2-bromo-3-chloropyridine, abbypharma ap-18-5389 | |
| C1=CC(=C(N=C1)Br)Cl | |
| 192.44 | |
| 192.44 | |
| Crystalline Powder |
| 60°C | |
| C5H3BrClN | |
| 5 g | |
| GOHBBINNYAWQGO-UHFFFAOYSA-N | |
| 2-bromo-3-chloropyridine | |
| 7016326 | |
| ≥98.0% (GC) | |
| 2-Bromo-3-chloropyridine |
Safety and Handling
TSCA : No