missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-chloropyridine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 96424-68-9
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
InChI Key GOHBBINNYAWQGO-UHFFFAOYSA-N
Synonym pyridine, 2-bromo-3-chloro, 3-chloro-2-bromopyridine, 2-bromo-3-chloro-pyridine, chloro bromo pyridine, pubchem5776, 2-bromo-3-pyridyl chloride, ksc486m1b, 2-bromo-3-chloropyridine, abbypharma ap-18-5389
PubChem CID 7016326
IUPAC Name 2-bromo-3-chloropyridine
SMILES C1=CC(=C(N=C1)Br)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC442810050
SDS View Documents
Thermo Scientific Chemicals
442810050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

96424-68-9
192.44
pyridine, 2-bromo-3-chloro, 3-chloro-2-bromopyridine, 2-bromo-3-chloro-pyridine, chloro bromo pyridine, pubchem5776, 2-bromo-3-pyridyl chloride, ksc486m1b, 2-bromo-3-chloropyridine, abbypharma ap-18-5389
2-bromo-3-chloropyridine
C5H3BrClN
GOHBBINNYAWQGO-UHFFFAOYSA-N
7016326
C1=CC(=C(N=C1)Br)Cl

Specifications

96424-68-9
C5H3BrClN
pyridine, 2-bromo-3-chloro, 3-chloro-2-bromopyridine, 2-bromo-3-chloro-pyridine, chloro bromo pyridine, pubchem5776, 2-bromo-3-pyridyl chloride, ksc486m1b, 2-bromo-3-chloropyridine, abbypharma ap-18-5389
C1=CC(=C(N=C1)Br)Cl
192.44
192.44
2-Bromo-3-chloropyridine
96% min. (GC)
5g
GOHBBINNYAWQGO-UHFFFAOYSA-N
2-bromo-3-chloropyridine
7016326
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only