missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-chloropyridine, 97%

CAS: 96424-68-9 | C5H3BrClN | 192.44 g/mol

$45.37 - $152.18

Chemical Identifiers

CAS 96424-68-9
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD04039344
InChI Key GOHBBINNYAWQGO-UHFFFAOYSA-N
Synonym pyridine, 2-bromo-3-chloro, 3-chloro-2-bromopyridine, 2-bromo-3-chloro-pyridine, chloro bromo pyridine, pubchem5776, 2-bromo-3-pyridyl chloride, ksc486m1b, 2-bromo-3-chloropyridine, abbypharma ap-18-5389
PubChem CID 7016326
IUPAC Name 2-bromo-3-chloropyridine
SMILES C1=CC(=C(N=C1)Br)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL2001203
View Documents
Thermo Scientific Chemicals
L2001203
1 g
Each for $45.37
Only null left
 
AAL2001206
View Documents
Thermo Scientific Chemicals
L2001206
5 g
Each for $152.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

96424-68-9
192.44
GOHBBINNYAWQGO-UHFFFAOYSA-N
7016326
C1=CC(=C(N=C1)Br)Cl
C5H3BrClN
MFCD04039344
pyridine, 2-bromo-3-chloro, 3-chloro-2-bromopyridine, 2-bromo-3-chloro-pyridine, chloro bromo pyridine, pubchem5776, 2-bromo-3-pyridyl chloride, ksc486m1b, 2-bromo-3-chloropyridine, abbypharma ap-18-5389
2-bromo-3-chloropyridine

Specifications

96424-68-9
C5H3BrClN
1 g
pyridine, 2-bromo-3-chloro, 3-chloro-2-bromopyridine, 2-bromo-3-chloro-pyridine, chloro bromo pyridine, pubchem5776, 2-bromo-3-pyridyl chloride, ksc486m1b, 2-bromo-3-chloropyridine, abbypharma ap-18-5389
C1=CC(=C(N=C1)Br)Cl
192.44
192.44
2-Bromo-3-chloropyridine
59°C to 61°C
MFCD04039344
1561638
GOHBBINNYAWQGO-UHFFFAOYSA-N
2-bromo-3-chloropyridine
7016326
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only