missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-chloropyridine, 97%

Catalog No. AAL2001206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2001206 5 g
AAL2001203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2001206 Supplier Thermo Scientific Chemicals Supplier No. L2001206

CAS: 96424-68-9 | C5H3BrClN | 192.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96424-68-9
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD04039344
InChI Key GOHBBINNYAWQGO-UHFFFAOYSA-N
Synonym pyridine, 2-bromo-3-chloro, 3-chloro-2-bromopyridine, 2-bromo-3-chloro-pyridine, chloro bromo pyridine, pubchem5776, 2-bromo-3-pyridyl chloride, ksc486m1b, 2-bromo-3-chloropyridine, abbypharma ap-18-5389
PubChem CID 7016326
IUPAC Name 2-bromo-3-chloropyridine
SMILES C1=CC(=C(N=C1)Br)Cl

Specifications

Melting Point 59°C to 61°C
Quantity 5 g
Beilstein 1561638
Formula Weight 192.44
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-chloropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only