missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(chloromethyl)pyridine, 96%, Thermo Scientific™

Catalog No. p-7034624
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5231303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5231303 Supplier Thermo Scientific Chemicals Supplier No. H5231303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 944906-87-0
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD10697618
InChI Key OWRYEOOYOHXZJX-UHFFFAOYSA-N
Synonym 2-bromo-3-chloromethyl pyridine, pyridine, 2-bromo-3-chloromethyl, 2-bromo-3-chloromethylene pyridine
PubChem CID 53416632
IUPAC Name 2-bromo-3-(chloromethyl)pyridine
SMILES C1=CC(=C(N=C1)Br)CCl

Specifications

Melting Point 34°C to 36°C
Quantity 1 g
Beilstein 15651009
Formula Weight 206.47
Percent Purity 96%
Chemical Name or Material 2-Bromo-3-(chloromethyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen