missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine 96.0+%, TCI America™
$68.97 - $225.56
Chemical Identifiers
| CAS | 75806-84-7 |
|---|---|
| Molecular Formula | C6H2BrClF3N |
| Molecular Weight (g/mol) | 260.438 |
| MDL Number | MFCD00153072 |
| InChI Key | SMTKGMYGLYWNDL-UHFFFAOYSA-N |
| Synonym | 2-bromo-3-chloro-5-trifluoromethyl pyridine, librarion l938, abbypharma ap-13-5059, pyridine, 2-bromo-3-chloro-5-trifluoromethyl, pubchem3000, acmc-1bicf, ksc498c5d, buttpark 153\33-66, 2-bromo-3-chloro-5 trifluoromethyl pyridine |
| PubChem CID | 2736237 |
| IUPAC Name | 2-bromo-3-chloro-5-(trifluoromethyl)pyridine |
| SMILES | C1=C(C=NC(=C1Cl)Br)C(F)(F)F |
Chemical Identifiers
| 75806-84-7 | |
| 260.438 | |
| SMTKGMYGLYWNDL-UHFFFAOYSA-N | |
| 2736237 | |
| C1=C(C=NC(=C1Cl)Br)C(F)(F)F |
| C6H2BrClF3N | |
| MFCD00153072 | |
| 2-bromo-3-chloro-5-trifluoromethyl pyridine, librarion l938, abbypharma ap-13-5059, pyridine, 2-bromo-3-chloro-5-trifluoromethyl, pubchem3000, acmc-1bicf, ksc498c5d, buttpark 153\33-66, 2-bromo-3-chloro-5 trifluoromethyl pyridine | |
| 2-bromo-3-chloro-5-(trifluoromethyl)pyridine |
Specifications
| 75806-84-7 | |
| C6H2BrClF3N | |
| 1 g | |
| SMTKGMYGLYWNDL-UHFFFAOYSA-N | |
| 2-bromo-3-chloro-5-(trifluoromethyl)pyridine | |
| 2736237 | |
| ≥96.0% (GC) | |
| 2-Bromo-3-chloro-5-(trifluoromethyl)pyridine |
| 80°C | |
| MFCD00153072 | |
| 2-bromo-3-chloro-5-trifluoromethyl pyridine, librarion l938, abbypharma ap-13-5059, pyridine, 2-bromo-3-chloro-5-trifluoromethyl, pubchem3000, acmc-1bicf, ksc498c5d, buttpark 153\33-66, 2-bromo-3-chloro-5 trifluoromethyl pyridine | |
| C1=C(C=NC(=C1Cl)Br)C(F)(F)F | |
| 260.438 | |
| 260.44 | |
| Liquid |
Safety and Handling
TSCA : No