missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3,3,3-trifluoro-1-propene 97.0+%, TCI America™

Catalog No. b180125g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B180125G 25 g
B18015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B180125G Supplier TCI America Supplier No. B180125G
Only null left

Chemical Identifiers

CAS 1514-82-5
Molecular Formula C3H2BrF3
Molecular Weight (g/mol) 174.95
MDL Number MFCD00077469
InChI Key QKBKGNDTLQFSEU-UHFFFAOYSA-N
Synonym 2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropene
PubChem CID 272696
IUPAC Name 2-bromo-3,3,3-trifluoroprop-1-ene
SMILES FC(F)(F)C(Br)=C

Specifications

Color Yellow
Boiling Point 34°C
Quantity 25 g
Formula Weight 174.95
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3,3,3-trifluoro-1-propene
TSCA No
Recommended Storage Refrigerator