missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(2-ethylhexyl)-5-iodothiophene (stabilized with Copper chip) 97.0+%, TCI America™

Catalog No. p-7123630
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45851G 1 g
B45855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45851G Supplier TCI America Supplier No. B45851G

Chemical Identifiers

CAS 1034352-30-1
Molecular Formula C12H18BrIS
Molecular Weight (g/mol) 401.144
InChI Key VHAVRRSIFMAOTI-UHFFFAOYSA-N
PubChem CID 70976734
IUPAC Name 2-bromo-3-(2-ethylhexyl)-5-iodothiophene
SMILES CCCCC(CC)CC1=C(SC(=C1)I)Br

Specifications

Quantity 1 g
Formula Weight 401.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-(2-ethylhexyl)-5-iodothiophene (stabilized with Copper chip)
TSCA No
Recommended Storage Freezer