missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-phenylacetophenone 95.0+%, TCI America™

Catalog No. p-7123562
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B44715G 5 g
B447125G 25 g
2 options

Catalog No. B44715G

Supplier: TCI America B44715G

Only null left

Chemical Identifiers

CAS 1484-50-0
Molecular Formula C14H11BrO
Molecular Weight (g/mol) 275.145
MDL Number MFCD00000136
InChI Key ZFFBIQMNKOJDJE-UHFFFAOYSA-N
PubChem CID 102630
IUPAC Name 2-bromo-1,2-diphenylethanone
SMILES C1=CC=C(C=C1)C(C(=O)C2=CC=CC=C2)Br

Specifications

Melting Point 56°C
Quantity 5 g
UN Number 3261
Synonym 2-bromo-2-phenylacetophenone, desyl bromide, 2-bromo-1,2-diphenylethan-1-one, ethanone, 2-bromo-1,2-diphenyl, ethanone,2-bromo-1,2-diphenyl, 2-bromo-1,2-diphenyl-ethanone, desylbromide, alpha-bromodeoxybenzoin, alpha-bromodesoxibenzoine
Formula Weight 275.15
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-2-phenylacetophenone